siRNA Details
| Gene Symbol: Sec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153125.2 | NP_694765.2 | 5 | 4077 | ||
| GenBank mRNA | CK638146.1 | - | 2 | 37 | ||
| CV883992.1 | - | 4 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec16a| Gene Symbol: Sec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153125.2 | NP_694765.2 | 5 | 4077 | ||
| GenBank mRNA | CK638146.1 | - | 2 | 37 | ||
| CV883992.1 | - | 4 | 355 | |||