siRNA Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290503.1 | NP_001277432.1 | 5 | 1210 | ||
| NM_178694.4 | NP_848809.2 | 5 | 1249 | |||
| GenBank mRNA | AK029290.1 | - | 5 | 1184 | ||
| AK032153.1 | - | 5 | 1180 | |||
| AK032318.1 | - | 4 | 856 | |||
| AK035712.1 | - | 2 | 162 | |||
| AK048853.1 | - | 5 | 1105 | |||
| AK081355.1 | - | 5 | 1087 | |||
| AK087046.1 | - | 5 | 1083 | |||
| AK155401.1 | - | 5 | 1157 | |||
| BC048924.1 | - | 5 | 1092 | 6312691 | ||
| BC057021.1 | - | 5 | 1154 | 6825937 | ||

Zer1
Genomic Map