siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198304.2 | NP_938046.2 | 29 | 3149 | ||
| GenBank mRNA | AK129073.1 | - | 29 | 3149 | ||
| BC032090.1 | - | 29 | 3151 | |||
| BC056489.1 | - | 15 | 1728 | |||
| BC096591.1 | - | 29 | 3150 | 4916413 | ||

Nup188
Genomic Map