siRNA Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290625.1 | NP_001277554.1 | 12 | 1639 | ||
| NM_001290626.1 | NP_001277555.1 | 11 | 1637 | |||
| NM_177345.4 | NP_796319.1 | 12 | 1965 | |||
| GenBank mRNA | AK027932.1 | - | 7 | 738 | ||
| AK052045.1 | - | 12 | 1633 | |||
| AK132263.1 | - | 12 | 1629 | |||
| BC043296.1 | - | 12 | 1864 | |||
| BC090644.1 | - | 11 | 1539 | 30629263 | ||
| BC096618.1 | - | 12 | 1938 | 6392317 | ||

Mapkap1
Genomic Map