siRNA Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145522.4 | NP_663497.2 | 6, 7 | 836 | ||
GenBank mRNA | AK035343.1 | - | 6, 7 | 716 | ||
BC019800.1 | - | 6, 7 | 712 | 5150243 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145522.4 | NP_663497.2 | 6, 7 | 836 | ||
GenBank mRNA | AK035343.1 | - | 6, 7 | 716 | ||
BC019800.1 | - | 6, 7 | 712 | 5150243 |