siRNA Details
| Gene Symbol: Epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172663.4 | NP_766251.3 | 9 | 1567 | ||
| GenBank mRNA | AK031025.1 | - | 9 | 1540 | ||
| AK047947.1 | - | 9 | 1235 | |||
| AK141073.1 | - | 858 | ||||
| AK164886.1 | - | 5 | 594 | |||
| BC048785.1 | - | 8 | 1006 | |||

Epc2
Genomic Map