siRNA Details
| Gene Symbol: Ccdc148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001178.1 | NP_001001178.1 | 651 | |||
| GenBank mRNA | BC062650.1 | - | 651 | 6417188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc148| Gene Symbol: Ccdc148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001178.1 | NP_001001178.1 | 651 | |||
| GenBank mRNA | BC062650.1 | - | 651 | 6417188 | ||