siRNA Details
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172664.3 | NP_766252.2 | 15, 16 | 1741 | ||
| GenBank mRNA | AK029773.1 | - | 14, 15 | 1467 | ||
| AK172897.1 | - | 9, 10 | 1005 | |||
| BC051641.1 | - | 15, 16 | 1741 | 6507968 | ||
| BC057369.1 | - | 17, 18 | 1980 | |||

Tlk1
Genomic Map