siRNA Details
| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172666.3 | NP_766254.2 | 3 | 493 | ||
| GenBank mRNA | AK031049.1 | - | 3 | 495 | ||
| AK031154.1 | - | 3 | 334 | |||
| AK042745.1 | - | 3 | 333 | |||
| AK053686.1 | - | 3 | 482 | |||
| AK079447.1 | - | 2 | 173 | |||
| BC063086.1 | - | 3 | 478 | 6332574 | ||
| BY713473.1 | - | 3 | 493 | |||

Agps
Genomic Map