siRNA Details
| Gene Symbol: Tnks1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081260.2 | NP_001074729.1 | 3 | 685 | ||
| GenBank mRNA | BC150947.1 | - | 3 | 605 | 9087858 | |
| CB723061.1 | - | 2 | 553 | |||
| CF733903.1 | - | 1 | 85 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnks1bp1| Gene Symbol: Tnks1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081260.2 | NP_001074729.1 | 3 | 685 | ||
| GenBank mRNA | BC150947.1 | - | 3 | 605 | 9087858 | |
| CB723061.1 | - | 2 | 553 | |||
| CF733903.1 | - | 1 | 85 | |||