siRNA Details
| Gene Symbol: 4833423E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081664.2 | NP_001075133.1 | 8 | 1261 | ||
| GenBank mRNA | AK076485.1 | - | 8 | 1350 | ||
| BC120879.2 | - | 8 | 1261 | 40057526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833423E24Rik| Gene Symbol: 4833423E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081664.2 | NP_001075133.1 | 8 | 1261 | ||
| GenBank mRNA | AK076485.1 | - | 8 | 1350 | ||
| BC120879.2 | - | 8 | 1261 | 40057526 | ||