siRNA Details
| Gene Symbol: Slc35c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145832.3 | NP_665831.1 | 2 | 479 | ||
| NM_211358.2 | NP_997597.1 | 1 | 967 | |||
| GenBank mRNA | AJ440720.1 | - | 1 | 364 | ||
| AK030977.1 | - | 2 | 480 | |||
| AK033295.1 | - | 1 | 56 | |||
| AK040986.1 | - | 1 | 714 | |||
| AK045972.1 | - | 2 | 456 | |||
| AK050311.1 | - | 2 | 444 | |||
| AK081068.1 | - | 1 | 465 | |||
| AK088227.1 | - | 2 | 466 | |||
| AK138595.1 | - | 1 | 910 | |||
| AK144762.1 | - | 1 | 889 | |||
| AK151154.1 | - | 1 | 963 | |||
| BC031814.1 | - | 1 | 561 | 4196818 | ||
| BC116736.1 | - | 1 | 392 | 40126055 | ||
| BC119128.1 | - | 1 | 392 | 8733877 | ||

Slc35c1
Genomic Map