siRNA Details
| Gene Symbol: Itpka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146125.2 | NP_666237.1 | 3, 4 | 835 | ||
| GenBank mRNA | BC027291.1 | - | 3, 4 | 835 | 3481738 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpka| Gene Symbol: Itpka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146125.2 | NP_666237.1 | 3, 4 | 835 | ||
| GenBank mRNA | BC027291.1 | - | 3, 4 | 835 | 3481738 | |