siRNA Details
| Gene Symbol: 4930402H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029432.1 | NP_083708.1 | 30 | 2828 | ||
| GenBank mRNA | AK133294.1 | - | 30 | 2970 | ||
| AK143611.1 | - | 30 | 2853 | |||
| BC052447.1 | - | 30 | 2828 | 6408647 | ||

4930402H24Rik
Genomic Map