siRNA Details
| Gene Symbol: Kiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033298.3 | NP_001028470.1 | 12 | 1900 | ||
| GenBank mRNA | AK135486.1 | - | 12 | 1903 | ||
| AK178867.1 | - | 3 | 165 | |||
| AK218233.1 | - | 3 | 173 | |||
| BC150844.1 | - | 12 | 1945 | 9087755 | ||
| BC150845.1 | - | 12 | 1945 | 9087756 | ||

Kiz
Genomic Map