siRNA Details
| Gene Symbol: Rspo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040689.1 | NP_001035779.1 | 3 | 482 | ||
| GenBank mRNA | AK020904.1 | - | 3 | 483 | ||
| AK043484.1 | - | 3 | 484 | |||
| BC048707.1 | - | 3 | 456 | 6511472 | ||
| BC116367.1 | - | 3 | 445 | |||

Rspo4
Genomic Map