siRNA Details
| Gene Symbol: Mylk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081044.2 | NP_001074513.2 | 10 | 1710 | ||
| GenBank mRNA | BC019408.1 | - | 5 | 397 | ||
| BC158041.1 | - | 10 | 1541 | 9088157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mylk2| Gene Symbol: Mylk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081044.2 | NP_001074513.2 | 10 | 1710 | ||
| GenBank mRNA | BC019408.1 | - | 5 | 397 | ||
| BC158041.1 | - | 10 | 1541 | 9088157 | ||