siRNA Details
| Gene Symbol: Tgif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291124.1 | NP_001278053.1 | 3 | 528 | ||
| NM_173396.3 | NP_775572.1 | 3 | 528 | |||
| GenBank mRNA | AK017884.1 | - | 3 | 396 | ||
| AK029490.1 | - | 3 | 458 | |||
| AK157623.1 | - | 3 | 436 | |||
| AK178732.1 | - | 2 | 142 | |||
| AK185386.1 | - | 2 | 162 | |||
| AK207133.1 | - | 2 | 142 | |||
| AK208156.1 | - | 2 | 142 | |||
| AK216126.1 | - | 2 | 142 | |||
| AK219241.1 | - | 2 | 142 | |||
| BC138705.1 | - | 3 | 441 | 8861727 | ||
| BC138706.1 | - | 3 | 443 | 8861728 | ||
| BC158022.1 | - | 2 | 290 | 9007328 | ||
| CF731285.1 | - | 2 | 146 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001134983, XM_006235424, XM_006235426, XM_006235425, XM_006235427, and others.

Tgif2
Genomic Map