siRNA Details
| Gene Symbol: Tgif2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291124.1 | NP_001278053.1 | 2 | 350 | ||
| NM_173396.3 | NP_775572.1 | 2 | 350 | |||
| GenBank mRNA | AK029490.1 | - | 2 | 280 | ||
| AK157623.1 | - | 2 | 258 | |||
| BC138705.1 | - | 2 | 263 | 8861727 | ||
| BC138706.1 | - | 2 | 265 | 8861728 | ||
| BC158022.1 | - | 1 | 112 | 9007328 | ||

Tgif2
Genomic Map