siRNA Details
| Gene Symbol: Zmynd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252584.2 | NP_001239513.1 | 12 | 1706 | ||
| NM_001252585.1 | NP_001239514.1 | 11 | 1598 | |||
| NM_001281926.1 | NP_001268855.1 | 12 | 1605 | |||
| NM_001291158.1 | NP_001278087.1 | 13 | 3715 | |||
| NM_027230.5 | NP_081506.3 | 13 | 1793 | |||
| GenBank mRNA | AB093284.2 | - | 13 | 3707 | ||
| AK132427.1 | - | 11 | 1600 | |||
| AK147620.1 | - | 12 | 1606 | |||
| AK155864.1 | - | 12 | 1511 | |||
| BC041797.1 | - | 2 | 521 | |||
| BC048186.1 | - | 13 | 1600 | 6827541 | ||

Zmynd8
Genomic Map