siRNA Details
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183161.3 | NP_898984.3 | 6 | 757 | ||
GenBank mRNA | AK006122.1 | - | 2 | 97 | ||
CF165170.1 | - | 3 | 243 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183161.3 | NP_898984.3 | 6 | 757 | ||
GenBank mRNA | AK006122.1 | - | 2 | 97 | ||
CF165170.1 | - | 3 | 243 |