siRNA Details
| Gene Symbol: Ythdf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173761.3 | NP_776122.1 | 4 | 583 | ||
| GenBank mRNA | AK033241.1 | - | 5 | 673 | ||
| AK046768.1 | - | 4 | 597 | |||
| AK075846.1 | - | 4 | 562 | |||
| AK172582.1 | - | 4 | 557 | |||
| BC061479.1 | - | 4 | 554 | 6334535 | ||
| BC065050.1 | - | 4 | 583 | 6418891 | ||

Ythdf1
Genomic Map