siRNA Details
| Gene Symbol: ADH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000668.5 | NP_000659.2 | 9 | 1795 | ||
| NM_001286650.1 | NP_001273579.1 | 10 | 1917 | |||
| GenBank mRNA | AF153821.1 | - | 9 | 1752 | ||
| BC033009.2 | - | 9 | 1754 | 4828235 | ||
| BX647469.1 | - | 9 | 3779 | |||
| D00137.1 | - | 9 | 1704 | |||
| FJ224285.1 | - | 9 | 1754 | |||
| M21692.1 | - | 9 | 1716 | |||
| X03350.1 | - | 9 | 1756 | |||

ADH1B
Genomic Map