siRNA Details
| Gene Symbol: Samd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172676.2 | NP_766264.2 | 5 | 1459 | ||
| GenBank mRNA | AK053048.1 | - | 5 | 1412 | ||
| AK053884.1 | - | 3 | 1040 | |||
| AK164204.1 | - | 5 | 1499 | |||
| AK171151.1 | - | 5 | 1465 | |||
| AK218993.1 | - | 1 | 99 | |||
| BC019475.1 | - | 5 | 1469 | |||
| BC052921.1 | - | 5 | 1459 | 6311715 | ||

Samd10
Genomic Map