siRNA Details
| Gene Symbol: Stoml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153156.1 | NP_694796.1 | 4 | 247 | ||
| GenBank mRNA | AB085692.1 | - | 4 | 247 | ||
| BC058224.1 | - | 293 | ||||
| BC080859.1 | - | 276 | ||||
| BC138667.1 | - | 4 | 328 | 8861688 | ||
| BC138668.1 | - | 4 | 328 | 8861689 | ||

Stoml3
Genomic Map