siRNA Details
| Gene Symbol: Spg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144987.1 | NP_001138459.1 | 7 | 1898 | ||
| NM_001144988.1 | NP_001138460.1 | 6 | 1511 | |||
| NM_144895.2 | NP_659144.1 | 7 | 1682 | |||
| GenBank mRNA | AK031545.1 | - | 6 | 1237 | ||
| AK035081.1 | - | 5 | 1370 | |||
| AK036569.1 | - | 5 | 834 | |||
| AK087388.1 | - | 7 | 1651 | |||
| AK089946.1 | - | 7 | 1683 | |||
| AK129171.1 | - | 6 | 1490 | |||
| AK147760.1 | - | 7 | 1899 | |||
| AK159951.1 | - | 7 | 1683 | |||
| BC022921.1 | - | 7 | 1650 | 4220506 | ||

Spg20
Genomic Map