siRNA Details
Gene Symbol: Gpr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173398.3 | NP_775574.1 | 2 | 435 | ||
GenBank mRNA | AK041381.1 | - | 2 | 407 | ||
AK041582.1 | - | 2 | 431 | |||
AK079674.1 | - | 2 | 436 | |||
AK083323.1 | - | 2 | 432 | |||
AK089287.1 | - | 2 | 388 | |||
AK154725.1 | - | 2 | 433 | |||
BC024054.1 | - | 3 | 361 | 5068166 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
