siRNA Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177662.2 | NP_808330.1 | 6, 7 | 868 | ||
| GenBank mRNA | AK034490.1 | - | 6, 7 | 868 | ||
| AK049470.1 | - | 6, 7 | 841 | |||
| AK143683.1 | - | 6, 7 | 884 | |||
| AK165930.1 | - | 6, 7 | 830 | |||
| AK166103.1 | - | 6, 7 | 837 | |||
| BC044664.1 | - | 6, 7 | 760 | |||
| BC098219.1 | - | 6, 7 | 842 | 1066724 | ||

Ctso
Genomic Map