siRNA Details
| Gene Symbol: Ctso | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177662.2 | NP_808330.1 | 2 | 184 | ||
| GenBank mRNA | AK034490.1 | - | 2 | 184 | ||
| AK040071.1 | - | 2 | 158 | |||
| AK049470.1 | - | 2 | 157 | |||
| AK143683.1 | - | 2 | 200 | |||
| AK165930.1 | - | 2 | 146 | |||
| AK166103.1 | - | 2 | 153 | |||
| BC044664.1 | - | 2 | 76 | |||
| BC098219.1 | - | 2 | 158 | 1066724 | ||

Ctso
Genomic Map