siRNA Details
| Gene Symbol: D930015E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172681.4 | NP_766269.3 | 11 | 1088 | ||
| GenBank mRNA | AK052409.1 | - | 11 | 1067 | ||
| AK086232.1 | - | 11 | 1105 | |||
| AK129240.1 | - | 5 | 459 | |||
| AK137619.1 | - | 11 | 1063 | |||
| AK154818.1 | - | 11 | 1089 | |||
| AY662647.1 | - | 11 | 1015 | |||
| BC062940.1 | - | 11 | 1064 | 6842400 | ||

D930015E06Rik
Genomic Map