siRNA Details
| Gene Symbol: Fhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033301.4 | NP_001028473.2 | 12 | 2504 | ||
| NM_001205355.1 | NP_001192284.1 | 12 | 2575 | |||
| GenBank mRNA | AK031946.1 | - | 11 | 1977 | ||
| AK145217.1 | - | 12 | 2495 | |||
| AK173244.1 | - | 12 | 2575 | |||
| BC137632.1 | - | 12 | 2544 | 8860651 | ||

Fhdc1
Genomic Map