siRNA Details
| Gene Symbol: ADH1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000668.5 | NP_000659.2 | 9 | 2155 | ||
| NM_001286650.1 | NP_001273579.1 | 10 | 2277 | |||
| GenBank mRNA | AF153821.1 | - | 9 | 2112 | ||
| D00137.1 | - | 9 | 2064 | |||
| FJ224285.1 | - | 9 | 2114 | |||
| M21692.1 | - | 9 | 2077 | |||
| X03350.1 | - | 9 | 2114 | |||

ADH1B
Genomic Map