siRNA Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144898.2 | NP_659147.2 | 10 | 1080 | ||
| GenBank mRNA | AK134001.1 | - | 537 | |||
| AK139708.1 | - | 1059 | ||||
| AK163280.1 | - | 10 | 940 | |||
| AK166236.1 | - | 10 | 1052 | |||
| AK166834.1 | - | 3 | 2604 | |||
| AK167586.1 | - | 10 | 1028 | |||
| AK170160.1 | - | 10 | 1063 | |||
| BC008103.1 | - | 10 | 1030 | 3966815 | ||
| BC108354.1 | - | 10 | 1014 | 5035560 | ||

Msto1
Genomic Map