siRNA Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144898.2 | NP_659147.2 | 9 | 922 | ||
| GenBank mRNA | AK134001.1 | - | 379 | |||
| AK139708.1 | - | 901 | ||||
| AK163280.1 | - | 9 | 782 | |||
| AK166236.1 | - | 9 | 894 | |||
| AK166834.1 | - | 2 | 2446 | |||
| AK167586.1 | - | 9 | 870 | |||
| AK170160.1 | - | 9 | 905 | |||
| AK177497.1 | - | 2 | 97 | |||
| BC008103.1 | - | 9 | 872 | 3966815 | ||
| BC108354.1 | - | 9 | 856 | 5035560 | ||

Msto1
Genomic Map