siRNA Details
| Gene Symbol: Ints3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145540.3 | NP_663515.2 | 9 | 1559 | ||
| NM_178876.3 | NP_849207.2 | 9 | 1559 | |||
| GenBank mRNA | AK033296.1 | - | 9 | 1226 | ||
| AK040147.1 | - | 9 | 1549 | |||
| AK146699.1 | - | 9 | 1453 | |||
| AK147865.1 | - | 9 | 1538 | |||
| AK212123.1 | - | 2 | 123 | |||
| BC055344.1 | - | 9 | 1094 | 6827687 | ||

Ints3
Genomic Map