siRNA Details
| Gene Symbol: Gm4858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034860.2 | NP_001030032.1 | 3 | 741 | ||
| NM_001177694.1 | NP_001171165.1 | 1 | 916 | |||
| GenBank mRNA | AK139723.1 | - | 3 | 742 | ||
| BC172161.1 | - | 1 | 900 | |||

Gm4858
Genomic Map