siRNA Details
| Gene Symbol: Golph3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177669.1 | NP_001171140.1 | 4 | 491 | ||
| NM_001177670.1 | NP_001171141.1 | 5 | 680 | |||
| NM_146133.3 | NP_666245.2 | 5 | 680 | |||
| GenBank mRNA | AK008434.1 | - | 5 | 647 | ||
| AK160454.1 | - | 4 | 452 | |||
| AK167675.1 | - | 5 | 630 | |||
| AK169724.1 | - | 4 | 519 | |||
| BC027194.1 | - | 5 | 590 | 4216692 | ||
| BC047147.1 | - | 5 | 655 | 6465983 | ||

Golph3l
Genomic Map