siRNA Details
| Gene Symbol: Trim45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165952.1 | NP_001159424.1 | 2 | 1520 | ||
| NM_001165953.1 | NP_001159425.1 | 2 | 1520 | |||
| NM_194343.2 | NP_919324.2 | 2 | 1520 | |||
| GenBank mRNA | AK076600.1 | - | 2 | 1096 | ||
| AK150574.1 | - | 2 | 1521 | |||
| AK153571.1 | - | 2 | 1521 | |||
| BC057358.1 | - | 2 | 1398 | 6835531 | ||

Trim45
Genomic Map