siRNA Details
Gene Symbol: Chil6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178412.2 | NP_848499.1 | 7 | 894 | ||
GenBank mRNA | BC051070.1 | - | 7 | 894 | 6308125 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chil6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178412.2 | NP_848499.1 | 7 | 894 | ||
GenBank mRNA | BC051070.1 | - | 7 | 894 | 6308125 |