siRNA Details
| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145542.3 | NP_663517.2 | 14 | 1627 | ||
| GenBank mRNA | AB092504.1 | - | 14 | 1356 | ||
| AK078823.1 | - | 7 | 829 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahcyl1| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145542.3 | NP_663517.2 | 14 | 1627 | ||
| GenBank mRNA | AB092504.1 | - | 14 | 1356 | ||
| AK078823.1 | - | 7 | 829 | |||