siRNA Details
| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145542.3 | NP_663517.2 | 2, 3 | 511 | ||
| GenBank mRNA | AB092504.1 | - | 2, 3 | 240 | ||
| AK087454.1 | - | 2, 3 | 469 | |||
| BC018218.1 | - | 2, 3 | 340 | 4007102 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahcyl1| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145542.3 | NP_663517.2 | 2, 3 | 511 | ||
| GenBank mRNA | AB092504.1 | - | 2, 3 | 240 | ||
| AK087454.1 | - | 2, 3 | 469 | |||
| BC018218.1 | - | 2, 3 | 340 | 4007102 | ||