siRNA Details
| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030016.2 | NP_084292.1 | 2 | 1701 | ||
| GenBank mRNA | AK020899.1 | - | 1 | 118 | ||
| AK039588.1 | - | 2 | 1702 | |||
| AK041636.1 | - | 1 | 167 | |||
| AK163561.1 | - | 1 | 116 | |||
| BC127174.1 | - | 1 | 59 | |||

Trmt13
Genomic Map