siRNA Details
| Gene Symbol: Znhit6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081094.1 | NP_001074563.1 | 9, 10 | 1502 | ||
| GenBank mRNA | AK010555.1 | - | 8, 9 | 847 | ||
| AK148729.1 | - | 9, 10 | 1479 | |||
| AK167835.1 | - | 10, 11 | 925 | |||
| AK168844.1 | - | 10, 11 | 918 | |||
| BC147498.1 | - | 9, 10 | 1388 | 9056360 | ||
| BC150966.1 | - | 9, 10 | 1388 | 9087878 | ||
| BC150967.1 | - | 9, 10 | 1388 | 9087879 | ||
| BC151198.1 | - | 9, 10 | 1388 | 9088086 | ||

Znhit6
Genomic Map