siRNA Details
| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173788.3 | NP_776149.1 | 12 | 2117 | ||
| GenBank mRNA | AK158195.1 | - | 7 | 668 | ||
| BC037458.1 | - | 7 | 573 | |||
| BC042470.1 | - | 12 | 2117 | 5137261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npr2| Gene Symbol: Npr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173788.3 | NP_776149.1 | 12 | 2117 | ||
| GenBank mRNA | AK158195.1 | - | 7 | 668 | ||
| BC037458.1 | - | 7 | 573 | |||
| BC042470.1 | - | 12 | 2117 | 5137261 | ||