siRNA Details
| Gene Symbol: Slc25a51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009949.3 | NP_001009949.3 | 3 | 943 | ||
| GenBank mRNA | AK051139.1 | - | 940 | |||
| AK146788.1 | - | 3 | 905 | |||
| AK150362.1 | - | 3 | 901 | |||
| AK151688.1 | - | 3 | 901 | |||
| AK152171.1 | - | 3 | 901 | |||
| AK156168.1 | - | 3 | 901 | |||
| AK156342.1 | - | 3 | 902 | |||
| AK164726.1 | - | 3 | 925 | |||
| AK165253.1 | - | 3 | 939 | |||
| BC089000.1 | - | 2 | 838 | 6418170 | ||

Slc25a51
Genomic Map