siRNA Details
| Gene Symbol: Fam206a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081420.1 | NP_001074889.1 | 1, 2 | 395 | ||
| GenBank mRNA | AK052647.1 | - | 1, 2 | 210 | ||
| BC026590.1 | - | 1, 2 | 374 | 4973099 | ||
| BC048548.1 | - | 1, 2 | 346 | 6590165 | ||

Fam206a
Genomic Map