siRNA Details
| Gene Symbol: Fam206a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081420.1 | NP_001074889.1 | 3 | 564 | ||
| GenBank mRNA | AK043588.1 | - | 2 | 1417 | ||
| AK052647.1 | - | 3 | 379 | |||
| AK164596.1 | - | 3 | 377 | |||
| BC026590.1 | - | 3 | 543 | 4973099 | ||
| BC048548.1 | - | 3 | 515 | 6590165 | ||
| BC062190.1 | - | 4 | 545 | |||

Fam206a
Genomic Map