siRNA Details
| Gene Symbol: Fam206a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081420.1 | NP_001074889.1 | 1 | 365 | ||
| GenBank mRNA | AK052647.1 | - | 1 | 180 | ||
| AK164596.1 | - | 1 | 90 | |||
| BC026590.1 | - | 1 | 344 | 4973099 | ||
| BC048548.1 | - | 1 | 316 | 6590165 | ||
| BC062190.1 | - | 2 | 258 | |||

Fam206a
Genomic Map