siRNA Details
| Gene Symbol: AI314180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172381.2 | NP_759013.2 | 18 | 2093 | ||
| GenBank mRNA | AK038775.1 | - | 19 | 2007 | ||
| AK043020.1 | - | 19 | 2010 | |||
| AK129127.1 | - | 1 | 33 | |||
| BC058684.1 | - | 18 | 2093 | 6834912 | ||

AI314180
Genomic Map