siRNA Details
| Gene Symbol: Haus6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173400.2 | NP_775576.2 | 16 | 2085 | ||
| GenBank mRNA | AK036635.1 | - | 7 | 750 | ||
| AK077681.1 | - | 16 | 2107 | |||
| AK136163.1 | - | 7 | 806 | |||
| AK169222.1 | - | 16 | 2087 | |||
| AK173206.1 | - | 15 | 3511 | |||
| BC064725.1 | - | 11 | 1282 | |||
| BC068237.1 | - | 16 | 2085 | 30550558 | ||

Haus6
Genomic Map